Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dhs-3
Entrez Gene ID
172603
Gene Name
protein-coding
Gene Aliases
CELE_T02E1.5
Chromosome Location
Chr.I: 8212359 - 8218361 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129036.3 NP_001122508.1 4 - 5 476 94
NM_001129037.4 NP_001122509.1 3 - 4 471 94

View Full Product Details