Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gla-3
Entrez Gene ID
172604
Gene Name
protein-coding
Gene Aliases
CELE_T02E1.3
Chromosome Location
Chr.I: 8214376 - 8217999 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001263777.3 NP_001250706.1 5 - 6 692 79
NM_059837.6 NP_492238.2 5 - 6 692 79
NM_059838.3 NP_492239.1 5 - 6 763 79

View Full Product Details