Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
spe-12
Entrez Gene ID
172606
Gene Name
Uncharacterized protein
Gene Aliases
CELE_T02E1.1
Chromosome Location
Chr.I: 8224539 - 8226297 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059841.2 NP_492242.1 4 - 5 490 113

View Full Product Details