Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
col-63
Entrez Gene ID
172607
Gene Name
Col_cuticle_N domain-containing protein
Gene Aliases
CELE_ZK265.2
Chromosome Location
Chr.I: 8243694 - 8244963 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059844.8 NP_492245.1 1 - 2 84 148

View Full Product Details