Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sre-23
Entrez Gene ID
172608
Gene Name
Serpentine Receptor, class E (epsilon)
Gene Aliases
CELE_ZK265.5
Chromosome Location
Chr.I: 8256301 - 8258939 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059846.3 NP_492247.2 7 - 8 888 124

View Full Product Details