Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cep-1
Entrez Gene ID
172616
Gene Name
protein-coding
Gene Aliases
CELE_F52B5.5
Chromosome Location
Chr.I: 8323910 - 8328859 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026307.7 NP_001021478.1 6 - 7 801 88
NM_001383463.1 5 - 6 475 88
GenBank mRNA AF440800.2 - 5 - 6 481 88

View Full Product Details