Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tba-5
Entrez Gene ID
184569
Gene Name
Tubulin alpha chain
Gene Aliases
CELE_F16D3.1
Chromosome Location
Chr.I: 8353382 - 8358273 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059867.3 NP_492268.1 4 - 5 860 68

View Full Product Details