Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ser-5
Entrez Gene ID
184572
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_F16D3.7
Chromosome Location
Chr.I: 8384931 - 8387039 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059872.3 NP_492273.2 3 - 4 672 64

View Full Product Details