Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
zhp-3
Entrez Gene ID
172644
Gene Name
protein-coding
Gene Aliases
CELE_K02B12.8
Chromosome Location
Chr.I: 8529372 - 8531342 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001263872.2 NP_001250801.1 6 - 7 949 68
NM_001263873.4 NP_001250802.1 6 - 7 961 68

View Full Product Details