Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gpa-15
Entrez Gene ID
172645
Gene Name
Guanine nucleotide-binding protein alpha-15 subunit
Gene Aliases
CELE_M04C7.1
Chromosome Location
Chr.I: 8531721 - 8535315 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059911.2 NP_492312.1 3 - 4 438 113
GenBank mRNA AY008137.1 - 2 - 3 307 113

View Full Product Details