Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rskn-1
Entrez Gene ID
172647
Gene Name
protein-coding
Gene Aliases
CELE_T01H8.1
Chromosome Location
Chr.I: 8573852 - 8580205 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026431.3 NP_001021602.1 4 - 5 671 110
NM_001026432.4 NP_001021603.1 5 - 6 684 110
NM_001129032.4 NP_001122504.1 5 - 6 690 110
NM_001373180.3 NP_001359590.1 6 - 7 906 110
NM_059918.4 NP_492319.2 5 - 6 734 110

View Full Product Details