Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rskn-1
Entrez Gene ID
172647
Gene Name
protein-coding
Gene Aliases
CELE_T01H8.1
Chromosome Location
Chr.I: 8573852 - 8580205 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373180.3 NP_001359590.1 3 - 4 436 103

View Full Product Details