Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
edc-3
Entrez Gene ID
172653
Gene Name
DFDF domain-containing protein
Gene Aliases
CELE_R05D11.8
Chromosome Location
Chr.I: 8602125 - 8605169 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059927.6 NP_492328.1 6 - 7 977 68

View Full Product Details