Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pbs-7
Entrez Gene ID
172674
Gene Name
Proteasome subunit beta
Gene Aliases
CELE_F39H11.5
Chromosome Location
Chr.I: 8695205 - 8696376 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059953.5 NP_492354.1 3 - 4 401 81

View Full Product Details