Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pas-4
Entrez Gene ID
172679
Gene Name
Proteasome subunit alpha type-7
Gene Aliases
CELE_C36B1.4
Chromosome Location
Chr.I: 8727218 - 8728487 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059959.7 NP_492360.1 2 - 3 355 121

View Full Product Details