Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rpb-3
Entrez Gene ID
172680
Gene Name
RPOLD domain-containing protein
Gene Aliases
CELE_C36B1.3
Chromosome Location
Chr.I: 8728760 - 8730556 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059960.7 NP_492361.2 2 - 3 266 85

View Full Product Details