Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
prp-4
Entrez Gene ID
183250
Gene Name
WD_REPEATS_REGION domain-containing protein
Gene Aliases
CELE_C36B1.5
Chromosome Location
Chr.I: 8733413 - 8735717 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059962.4 NP_492363.1 6 - 7 954 132

View Full Product Details