Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lmn-1
Entrez Gene ID
172687
Gene Name
Lamin-1
Gene Aliases
CELE_DY3.2
Chromosome Location
Chr.I: 8761690 - 8765623 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059970.6 NP_492371.1 6 - 7 735 80

View Full Product Details