Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hlh-16
Entrez Gene ID
183973
Gene Name
BHLH domain-containing protein
Gene Aliases
CELE_DY3.3
Chromosome Location
Chr.I: 8766840 - 8768082 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059971.3 NP_492372.2 2 - 3 410 116

View Full Product Details