Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pqn-26
Entrez Gene ID
183974
Gene Name
Glutaredoxin domain-containing protein
Gene Aliases
CELE_DY3.5
Chromosome Location
Chr.I: 8771178 - 8777518 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059974.5 NP_492375.3 7 - 8 1678 97

View Full Product Details