Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sup-17
Entrez Gene ID
172689
Gene Name
Disintegrin and metalloproteinase domain-containing protein 10 homolog
Gene Aliases
CELE_DY3.7
Chromosome Location
Chr.I: 8792957 - 8797298 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059976.7 NP_492377.1 10 - 11 1414 93
GenBank mRNA AF024614.1 - 10 - 11 1405 93

View Full Product Details