Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cids-2
Entrez Gene ID
172691
Gene Name
protein-coding
Gene Aliases
CELE_F36A2.1
Chromosome Location
Chr.I: 8804799 - 8808081 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128999.4 NP_001122471.1 7 - 8 943 67
NM_001129000.5 NP_001122472.1 7 - 8 991 67
NM_059979.6 NP_492380.2 7 - 8 940 67
NM_182083.6 NP_871883.2 7 - 8 925 67
NR_023944.1 6 - 7 918 67

View Full Product Details