Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hum-1
Entrez Gene ID
172698
Gene Name
Heavy chain, Unconventional Myosin
Gene Aliases
CELE_F29D10.4
Chromosome Location
Chr.I: 8845809 - 8851116 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059992.4 NP_492393.1 4 - 5 568 128

View Full Product Details