Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- thoc-3
- Entrez Gene ID
-
172713
- Gene Name
- WD_REPEATS_REGION domain-containing protein
- Gene Aliases
-
CELE_F32H2.4
- Chromosome Location
-
Chr.I: 8970294 - 8973054 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_060015.5 |
NP_492416.2 |
9 - 10 |
1341 |
110 |
- Gene Symbol
- F32H2.10
- Entrez Gene ID
-
260106
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_F32H2.10
- Chromosome Location
-
Chr.I: 8970286 - 8972970 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_060014.5 |
NP_492415.1 |
8 - 9 |
1462 |
110 |
View Full Product Details