Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fcp-1
Entrez Gene ID
172719
Gene Name
RNA polymerase II subunit A C-terminal domain phosphatase
Gene Aliases
CELE_F36F2.6
Chromosome Location
Chr.I: 9004513 - 9008708 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060022.5 NP_492423.2 5 - 6 1030 115

View Full Product Details