Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tax-2
Entrez Gene ID
172723
Gene Name
Cyclic nucleotide-binding domain-containing protein
Gene Aliases
CELE_F36F2.5
Chromosome Location
Chr.I: 9020945 - 9025608 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060026.6 NP_492427.3 3 - 4 570 118

View Full Product Details