Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hgo-1
Entrez Gene ID
172726
Gene Name
Homogentisate 1,2-dioxygenase
Gene Aliases
CELE_W06D4.1
Chromosome Location
Chr.I: 9049460 - 9055457 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060032.7 NP_492433.1 8 - 9 1311 79

View Full Product Details