Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rad-54.L
Entrez Gene ID
172728
Gene Name
Uncharacterized protein
Gene Aliases
CELE_W06D4.6
Chromosome Location
Chr.I: 9065079 - 9070264 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060037.5 NP_492438.1 12 - 13 1749 101

View Full Product Details