Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gei-17
Entrez Gene ID
172733
Gene Name
protein-coding
Gene Aliases
CELE_W10D5.3
Chromosome Location
Chr.I: 9100845 - 9107848 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026506.5 NP_001021677.3 8 - 9 833 67
NM_001026507.5 NP_001021678.3 8 - 9 1085 67
NM_001026508.6 NP_001021679.3 7 - 8 633 67
NM_060041.6 NP_492442.3 8 - 9 819 67
NM_060042.5 NP_492443.3 8 - 9 805 67
NM_060043.5 NP_492444.4 8 - 9 984 67
NR_000051.1 9 - 10 1003 67

View Full Product Details