Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- nduf-7
- Entrez Gene ID
-
172734
- Gene Name
- Probable NADH dehydrogenase [ubiquinone] iron-sulfur protein 7, mitochondrial
- Gene Aliases
-
CELE_W10D5.2
- Chromosome Location
-
Chr.I: 9108173 - 9110929 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_060044.6 |
NP_492445.1 |
2 - 3 |
357 |
60 |
View Full Product Details