Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
aspm-1
Entrez Gene ID
172746
Gene Name
Calponin-homology (CH) domain-containing protein
Gene Aliases
CELE_C45G3.1
Chromosome Location
Chr.I: 9221750 - 9226390 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060066.5 NP_492467.1 1 - 2 67 94

View Full Product Details