Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lin-41
Entrez Gene ID
172760
Gene Name
protein-coding
Gene Aliases
CELE_C12C8.3
Chromosome Location
Chr.I: 9334800 - 9342496 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025827.5 NP_001020998.1 8 - 9 1451 63
NM_001025828.6 NP_001020999.1 8 - 9 1451 63
GenBank mRNA AF195610.1 - 7 - 8 1256 63
AF195611.1 - 7 - 8 1256 63

View Full Product Details