Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dyf-5
Entrez Gene ID
187442
Gene Name
protein-coding
Gene Aliases
CELE_M04C9.5
Chromosome Location
Chr.I: 9359067 - 9364238 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136314.5 NP_001129786.2 2 - 3 199 96
NM_060092.4 NP_492493.2 2 - 3 191 96
GenBank mRNA AM498742.1 - 2 - 3 191 96

View Full Product Details