The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001026296.3 | NP_001021467.1 | 5 - 6 | 1969 | 110 |
NM_001026297.6 | NP_001021468.1 | 5 - 6 | 844 | 110 |