Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
twk-29
Entrez Gene ID
185828
Gene Name
protein-coding
Gene Aliases
CELE_F46A9.3
Chromosome Location
Chr.I: 9463751 - 9468917 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026296.3 NP_001021467.1 7 - 8 2265 75
NM_001026297.6 NP_001021468.1 7 - 8 1140 75

View Full Product Details