Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
set-32
Entrez Gene ID
183391
Gene Name
SET domain-containing protein
Gene Aliases
CELE_C41G7.4
Chromosome Location
Chr.I: 9519244 - 9521275 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060128.3 NP_492529.2 6 - 7 1469 97

View Full Product Details