Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ahr-1
Entrez Gene ID
172788
Gene Name
protein-coding
Gene Aliases
CELE_C41G7.5
Chromosome Location
Chr.I: 9522550 - 9531132 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025865.4 NP_001021036.1 12 - 13 1696 79
NM_001025866.5 NP_001021037.1 11 - 12 1630 79
GenBank mRNA AF039570.1 - 12 - 13 1692 79

View Full Product Details