Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
npp-14
Entrez Gene ID
172813
Gene Name
Nuclear pore complex protein 14
Gene Aliases
CELE_C03D6.4
Chromosome Location
Chr.I: 9660174 - 9664915 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060167.7 NP_492568.1 6 - 7 3773 72

View Full Product Details