Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cel-1
Entrez Gene ID
172814
Gene Name
protein-coding
Gene Aliases
CELE_C03D6.3
Chromosome Location
Chr.I: 9665194 - 9668648 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025808.5 NP_001020979.1 11 - 12 1756 128
NM_001025809.2 NP_001020980.1 11 - 12 1698 128
GenBank mRNA AF003925.1 - 11 - 12 1603 128

View Full Product Details