Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fce-1
Entrez Gene ID
172820
Gene Name
CAAX prenyl protease 1 homolog
Gene Aliases
CELE_C04F12.10
Chromosome Location
Chr.I: 9711052 - 9712719 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060181.6 NP_492582.1 2 - 3 236 165

View Full Product Details