Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
inx-20
Entrez Gene ID
188818
Gene Name
protein-coding
Gene Aliases
CELE_T23H4.1
Chromosome Location
Chr.I: 9874698 - 9879218 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001264306.2 NP_001251235.1 10 - 11 1315 75
NM_001264307.3 NP_001251236.1 10 - 11 1270 75
NM_001264308.4 NP_001251237.1 9 - 10 1237 75

View Full Product Details