Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-69
Entrez Gene ID
172840
Gene Name
Nuclear hormone receptor family member nhr-69
Gene Aliases
CELE_T23H4.2
Chromosome Location
Chr.I: 9879183 - 9881504 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060214.5 NP_492615.2 5 - 6 685 160
GenBank mRNA AY204178.1 - 5 - 6 705 160

View Full Product Details