Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ztf-11
Entrez Gene ID
172845
Gene Name
protein-coding
Gene Aliases
CELE_F52F12.6
Chromosome Location
Chr.I: 9902910 - 9910545 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001264316.2 NP_001251245.1 2 - 3 364 68
NM_001264317.3 NP_001251246.1 1 - 2 55 68

View Full Product Details