Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sqv-5
Entrez Gene ID
172851
Gene Name
protein-coding
Gene Aliases
CELE_T24D1.1
Chromosome Location
Chr.I: 9947804 - 9953391 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026468.8 NP_001021639.3 5 - 6 1992 96
NM_001026469.8 NP_001021640.2 6 - 7 2412 96
NM_001313510.3 NP_001300439.1 6 - 7 2418 96
GenBank mRNA AB088397.1 - 5 - 6 1992 96

View Full Product Details