Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tsp-7
Entrez Gene ID
192062
Gene Name
Tetraspanin
Gene Aliases
CELE_T23D8.2
Chromosome Location
Chr.I: 9969972 - 9973177 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060235.2 NP_492636.1 6 - 7 677 57

View Full Product Details