Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sys-1
Entrez Gene ID
172859
Gene Name
protein-coding
Gene Aliases
CELE_T23D8.9
Chromosome Location
Chr.I: 9980063 - 9984794 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313509.3 NP_001300438.1 4 - 5 890 92
NM_060238.6 NP_492639.1 6 - 7 1297 92

View Full Product Details