Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fog-3
Entrez Gene ID
172863
Gene Name
Anti_prolifrtn domain-containing protein
Gene Aliases
CELE_C03C11.2
Chromosome Location
Chr.I: 10016765 - 10018121 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060245.6 NP_492646.1 1 - 2 85 81
GenBank mRNA AJ011777.1 - 1 - 2 100 81

View Full Product Details