Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-87
Entrez Gene ID
172885
Gene Name
C-type lectin domain-containing protein 87
Gene Aliases
CELE_C25A1.8
Chromosome Location
Chr.I: 10184478 - 10185520 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060281.5 NP_492682.1 1 - 2 224 71
GenBank mRNA DQ340627.1 - 1 - 2 150 71

View Full Product Details