Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lin-11
Entrez Gene ID
172893
Gene Name
Protein lin-11
Gene Aliases
CELE_ZC247.3
Chromosome Location
Chr.I: 10248227 - 10255327 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060295.5 NP_492696.1 8 - 9 1083 64

View Full Product Details