Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rps-7
Entrez Gene ID
172901
Gene Name
40S ribosomal protein S7
Gene Aliases
CELE_ZC434.2
Chromosome Location
Chr.I: 10330879 - 10331808 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060307.4 NP_492708.1 3 - 4 473 63

View Full Product Details